Assay Details
Gene Symbol: LOC498750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017510.3 | NP_001017510.1 | 1 | 171 | 144 | |
GenBank mRNA | AY325226.1 | - | 1 | 171 | 144 | |
BC085940.1 | - | 1 - 2 | 632 | 7123624 | 144 | |
BC089212.1 | - | 1 - 2 | 430 | 7302835 | 144 |
Gene Symbol: LOC100911664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751691.3 | XP_003751739.1 | 1 - 2 | 2347 | 144 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
