Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207610.1 | NP_997493.1 | 18 - 19 | 3061 | 81 | |
| XM_006242938.2 | XP_006243000.1 | 19 - 20 | 3275 | 81 | ||
| GenBank mRNA | AY325198.1 | - | 18 - 19 | 3013 | 81 | |
| BC061761.1 | - | 18 - 19 | 3061 | 5621078 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P800048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119642 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P800048 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P800048 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P800048 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P800048 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42147 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82042 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94483 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89287 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10916941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916933 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916931 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916934 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916945 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10916923 | - | - |
| Rat Genome 230 2.0 Array | 1384053_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385615_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372888_at | - | - |
| Rat Genome 230 2.0 Array | 1384053_at | - | - |
| Rat Genome 230 2.0 Array | 1380496_at | - | - |
| Rat Genome 230 2.0 Array | 1385615_at | - | - |
| Rat Genome 230 2.0 Array | 1372888_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380496_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380496_at | - | - |
| Affymetrix HT Rat Focus Array | 1384053_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372888_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385615_at | - | - |
| Affymetrix HT Rat Focus Array | 1380496_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384053_at | - | - |
| Affymetrix HT Rat Focus Array | 1385615_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372888_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800019484.rn.1 | TC0800002428.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800019483.rn.1 | TC0800002428.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800019485.rn.1 | TC0800002428.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372888_at | - | - |
| Rat Expression Set 230 | 1372888_PM_at | - | - |
| Rat Expression Set 230 | 1380496_at | - | - |
| Rat Expression Set 230 | 1385615_at | - | - |
| Rat Expression Set 230 | 1380496_PM_at | - | - |
| Rat Expression Set 230 | 1385615_PM_at | - | - |
| Rat Expression Set 230 | 1384053_PM_at | - | - |
| Rat Expression Set 230 | 1384053_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916923 | - | - |
| Rat Genome U34 Set | rc_AI105144_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22176262 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348961 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49300 | - | - |

Ube4a
Genomic Map