Assay Details
| Gene Symbol: Rexo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008326.1 | NP_001008327.1 | 4 - 5 | 572 | 117 | |
| XM_006243001.2 | XP_006243063.1 | 4 - 5 | 486 | 117 | ||
| GenBank mRNA | BC086420.1 | - | 4 - 5 | 572 | 7300740 | 117 |
More Information
Updated Mapping Information
Set Membership: |
|

Rexo2
Genomic Map