Assay Details
| Gene Symbol: Antxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044249.2 | NP_001037714.1 | 7 - 8 | 950 | 63 | |
| GenBank mRNA | AY754025.1 | - | 6 - 7 | 381 | 63 | |
| BC131853.1 | - | 7 - 8 | 950 | 7320366 | 63 | |
| DQ789143.1 | - | 7 - 8 | 599 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P127397 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P127392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18634 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127397 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127397 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127392 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P127392 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P127397 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21885 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83957 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2367 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26436 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34168 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46601 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102372 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47065 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78636 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82502 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81297 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56624 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63324 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11159 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10863781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863779 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863778 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863780 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10863777 | - | - |
| Rat Genome 230 2.0 Array | 1396109_at | - | - |
| Rat Genome 230 2.0 Array | 1396109_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400023210.rn.1 | TC0400003262.rn.1 | RTA TC |
| Rat Expression Set 230 | 1396109_PM_at | - | - |
| Rat Expression Set 230 | 1396109_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863777 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359770 | - | - |

Antxr1
Genomic Map