Assay Details
| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033064.1 | NP_001028236.1 | 1 - 2 | 110 | 75 | |
| GenBank mRNA | BC103652.1 | - | 1 - 2 | 110 | 7317353 | 75 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103652.1, NM_001033064.1 transcripts.
Set Membership: |
|

Kazald1
Genomic Map