Assay Details
| Gene Symbol: Rps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277243.1 | NP_001264172.1 | 3 - 4 | 413 | 81 | |
| NM_001277244.1 | NP_001264173.1 | 3 - 4 | 413 | 81 | ||
| GenBank mRNA | BC158718.1 | - | 3 - 4 | 371 | 9089281 | 81 |
| FM052146.1 | - | 3 - 4 | 369 | 81 | ||
| FQ209785.1 | - | 3 - 4 | 387 | 81 | ||
| FQ210357.1 | - | 3 - 4 | 387 | 81 | ||
| FQ210491.1 | - | 3 - 4 | 370 | 81 | ||
| FQ217060.1 | - | 3 - 4 | 387 | 81 | ||
| FQ219286.1 | - | 3 - 4 | 387 | 81 | ||
| FQ221071.1 | - | 3 - 4 | 387 | 81 | ||
| FQ221219.1 | - | 3 - 4 | 387 | 81 | ||
| FQ221255.1 | - | 3 - 4 | 387 | 81 | ||
| FQ221284.1 | - | 3 - 4 | 387 | 81 | ||
| FQ221608.1 | - | 3 - 4 | 81 | |||
| FQ222025.1 | - | 3 - 4 | 387 | 81 | ||
| FQ222269.1 | - | 3 - 4 | 387 | 81 | ||
| FQ222365.1 | - | 3 - 4 | 387 | 81 | ||
| FQ222894.1 | - | 3 - 4 | 387 | 81 | ||
| FQ223194.1 | - | 3 - 4 | 432 | 81 | ||
| FQ224029.1 | - | 3 - 4 | 387 | 81 | ||
| FQ224177.1 | - | 3 - 4 | 387 | 81 | ||
| FQ225463.1 | - | 3 - 4 | 387 | 81 | ||
| FQ226199.1 | - | 3 - 4 | 387 | 81 | ||
| FQ226257.1 | - | 3 - 4 | 390 | 81 | ||
| FQ226702.1 | - | 3 - 4 | 407 | 81 | ||
| FQ227598.1 | - | 3 - 4 | 387 | 81 | ||
| FQ228474.1 | - | 3 - 4 | 387 | 81 | ||
| FQ230232.1 | - | 3 - 4 | 386 | 81 | ||
| FQ232912.1 | - | 3 - 4 | 387 | 81 | ||
| X58465.1 | - | 3 - 4 | 387 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps5
Genomic Map