Assay Details
Gene Symbol: Hip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100475.1 | NP_001093945.1 | 8 - 9 | 808 | 58 | |
XM_006249098.2 | XP_006249160.1 | 8 - 9 | 896 | 58 | ||
XM_006249099.2 | XP_006249161.1 | 8 - 9 | 934 | 58 | ||
XM_006249100.2 | XP_006249162.1 | 8 - 9 | 826 | 58 | ||
XM_008769114.1 | XP_008767336.1 | 8 - 9 | 896 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
actin cytoskeleton organization
regulation of apoptotic process
positive regulation of receptor-mediated endocytosis
clathrin coat assembly
apoptotic signaling pathway
actin binding
clathrin binding
protein binding, bridging
phosphatidylinositol binding
endocytosis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
actin cytoskeleton organization
regulation of apoptotic process
positive regulation of receptor-mediated endocytosis
clathrin coat assembly
apoptotic signaling pathway
actin binding
clathrin binding
protein binding, bridging
phosphatidylinositol binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P503022 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029192 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P503022 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029192 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P503022 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029192 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P503022 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P250281 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10757671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757646 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757643 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757644 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757655 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10757669 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10757642 | - | - |
Rat Genome 230 2.0 Array | 1390082_at | - | - |
Rat Genome 230 2.0 Array | 1388193_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383890_at | - | - |
Rat Genome 230 2.0 Array | 1392941_at | - | - |
Rat Genome 230 2.0 Array | 1390082_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383890_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388193_at | - | - |
Rat Genome 230 2.0 Array | 1392941_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389132_at | - | - |
Rat Genome 230 2.0 Array | 1389132_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388193_at | - | - |
Affymetrix HT Rat Focus Array | 1392941_at | - | - |
Affymetrix HT Rat Focus Array | 1392941_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390082_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388193_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390082_at | - | - |
Affymetrix HT Rat Focus Array | 1383890_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383890_at | - | - |
Affymetrix HT Rat Focus Array | 1389132_at | - | - |
Affymetrix HT Rat Focus Array | 1389132_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200003728.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003730.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003729.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003733.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003727.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003734.rn.1 | TC1200000422.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200003735.rn.1 | TC1200000422.rn.1 | RTA TC |
Rat Expression Set 230 | 1392941_at | - | - |
Rat Expression Set 230 | 1389132_PM_at | - | - |
Rat Expression Set 230 | 1390082_at | - | - |
Rat Expression Set 230 | 1388193_at | - | - |
Rat Expression Set 230 | 1388193_PM_at | - | - |
Rat Expression Set 230 | 1389132_at | - | - |
Rat Expression Set 230 | 1392941_PM_at | - | - |
Rat Expression Set 230 | 1390082_PM_at | - | - |
Rat Expression Set 230 | 1383890_PM_at | - | - |
Rat Expression Set 230 | 1383890_at | - | - |
Rat Genome U34 Set | rc_AA859948_at | - | - |
Rat Genome U34 Set | rc_AI010493_at | - | - |
Rat Genome U34 Set | rc_AI112383_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21632602 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375389 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376352 | - | - |
