Assay Details
Gene Symbol: LOC102553065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257725.2 | XP_006257787.2 | 7 - 8 | 717 | 67 |
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053730.1 | NP_446182.1 | 23 - 24 | 2483 | 67 | |
XM_006249020.2 | XP_006249082.1 | 23 - 24 | 2651 | 67 | ||
GenBank mRNA | AY027880.1 | - | 23 - 24 | 2483 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
