Assay Details
| Gene Symbol: Ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106006.1 | NP_001099476.1 | 2 - 3 | 373 | 140 | |
| XM_006250934.2 | XP_006250996.1 | 2 - 3 | 570 | 140 | ||
| GenBank mRNA | FQ226880.1 | - | 2 - 3 | 365 | 140 | |
| FQ233155.1 | - | 2 - 3 | 321 | 140 | ||
| FQ234076.1 | - | 2 - 3 | 351 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube2k
Genomic Map