Assay Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033652.1 | NP_001028824.1 | 6 - 7 | 764 | 94 | |
| NM_022630.1 | NP_072152.1 | 1 - 2 | 135 | 94 | ||
| GenBank mRNA | AF198087.1 | - | 1 - 2 | 135 | 94 | |
| AF453776.1 | - | 6 - 7 | 764 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11d
Genomic Map