Assay Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127494.1 | NP_001120966.1 | 441 | 142 | ||
| GenBank mRNA | EU371958.1 | - | 441 | 142 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stk24
Genomic Map