Assay Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100505.1 | NP_001093975.1 | 2 - 3 | 319 | 107 | |
| XM_008772367.1 | XP_008770589.1 | 3 - 4 | 1140 | 107 | ||
| XM_008772368.1 | XP_008770590.1 | 3 - 4 | 1268 | 107 | ||
| XM_008772369.1 | XP_008770591.1 | 3 - 4 | 1263 | 107 | ||
| GenBank mRNA | BC098819.1 | - | 2 - 3 | 254 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asna1
Genomic Map