Assay Details
| Gene Symbol: Junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021836.2 | NP_068608.2 | 1 | 1001 | 99 | |
| GenBank mRNA | BC061862.1 | - | 1 | 1001 | 5597977 | 99 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Junb
Genomic Map