Assay Details
| Gene Symbol: Mgat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107068.1 | NP_001100538.1 | 11 - 12 | 1632 | 61 | |
| XM_006247799.2 | XP_006247861.1 | 10 - 11 | 1951 | 61 | ||
| XM_008768373.1 | XP_008766595.1 | 10 - 11 | 1952 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071178 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P455228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149367 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071178 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P455228 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071178 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P455228 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P455228 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071178 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P455228 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104191 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10739859 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739853 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739862 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739850 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739854 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739848 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739851 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739858 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739860 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739857 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739849 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739861 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739852 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739855 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739847 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10739845 | - | - |
| Rat Genome 230 2.0 Array | 1380731_at | - | - |
| Rat Genome 230 2.0 Array | 1380731_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380731_at | - | - |
| Affymetrix HT Rat Focus Array | 1380731_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000017948.rn.1 | TC1000001988.rn.1 | RTA TC |
| Rat Expression Set 230 | 1380731_PM_at | - | - |
| Rat Expression Set 230 | 1380731_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22269788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1651130 | - | - |

Mgat5b
Genomic Map