Assay Details
| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012920.1 | NP_037052.1 | 11 - 12 | 943 | 93 | |
| GenBank mRNA | J02942.1 | - | 11 - 12 | 943 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk2a
Genomic Map