Assay Details
| Gene Symbol: Fam175b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106307.1 | NP_001099777.2 | 1 - 2 | 94 | 58 | |
| GenBank mRNA | FQ218234.1 | - | 1 - 2 | 83 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam175b
Genomic Map