Assay Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017316.2 | NP_059012.2 | 5 - 6 | 689 | 63 | |
| XM_006235067.2 | XP_006235129.1 | 5 - 6 | 1486 | 63 | ||
| XM_006235068.2 | XP_006235130.1 | 6 - 7 | 1489 | 63 | ||
| XM_006235069.2 | XP_006235131.1 | 3 - 4 | 2931 | 63 | ||
| GenBank mRNA | AF080453.1 | - | 5 - 6 | 689 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a2
Genomic Map