Assay Details
| Gene Symbol: Hsd17b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212529.1 | NP_997694.1 | 1 - 2 | 65 | 94 | |
| XM_006255965.2 | XP_006256027.1 | 1 - 2 | 180 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_212529.1, XM_006255965.2 transcripts.
Set Membership: |
|

Hsd17b8
Genomic Map