Assay Details
| Gene Symbol: Rrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013236.1 | NP_001013254.1 | 13 - 14 | 1679 | 105 | |
| GenBank mRNA | BC085906.1 | - | 13 - 14 | 1679 | 7193834 | 105 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rrm1
Genomic Map