Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CA510739.1 | - | 2 - 3 | 254 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
