Assay Details
| Gene Symbol: Scube3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271362.1 | NP_001258291.1 | 4 - 5 | 473 | 66 | |
| XM_008772802.1 | XP_008771024.1 | 3 - 4 | 2438 | 66 | ||
| XM_008772803.1 | XP_008771025.1 | 4 - 5 | 495 | 66 | ||
| XM_008772804.1 | XP_008771026.1 | 3 - 4 | 2438 | 66 | ||
| XM_008772805.1 | XP_008771027.1 | 3 - 4 | 2438 | 66 | ||
| XM_008772806.1 | XP_008771028.1 | 3 - 4 | 2438 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scube3
Genomic Map