Assay Details
| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764735.1 | XP_008762957.1 | 1 - 2 | 136 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764735.1 transcripts.
Set Membership: |
|

Jdp2
Genomic Map