Assay Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759036.1 | XP_008757258.1 | 2 - 3 | 688 | 72 | |
| XM_008759037.1 | XP_008757259.1 | 2 - 3 | 688 | 72 | ||
| XM_008759038.1 | XP_008757260.1 | 2 - 3 | 688 | 72 | ||
| XM_008759039.1 | XP_008757261.1 | 2 - 3 | 688 | 72 | ||
| GenBank mRNA | BC160851.1 | - | 2 - 3 | 585 | 72 | |
More Information
Set Membership: |
|

Dmwd
Genomic Map