Assay Details
| Gene Symbol: Tpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053867.1 | NP_446319.1 | 2 - 3 | 168 | 96 | |
| GenBank mRNA | BC086358.1 | - | 2 - 3 | 179 | 7303675 | 96 |
| FQ210845.1 | - | 1 - 2 | 82 | 96 | ||
| FQ217312.1 | - | 2 - 3 | 196 | 96 | ||
| FQ217485.1 | - | 2 - 3 | 196 | 96 | ||
| FQ220662.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221184.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221191.1 | - | 2 - 3 | 185 | 96 | ||
| FQ221220.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221227.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221357.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221503.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221524.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221585.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221695.1 | - | 2 - 3 | 196 | 96 | ||
| FQ221696.1 | - | 2 - 3 | 197 | 96 | ||
| FQ222296.1 | - | 2 - 3 | 196 | 96 | ||
| FQ222704.1 | - | 2 - 3 | 195 | 96 | ||
| FQ222827.1 | - | 2 - 3 | 196 | 96 | ||
| FQ223078.1 | - | 2 - 3 | 196 | 96 | ||
| FQ223172.1 | - | 2 - 3 | 197 | 96 | ||
| FQ223294.1 | - | 2 - 3 | 203 | 96 | ||
| FQ223538.1 | - | 2 - 3 | 196 | 96 | ||
| FQ223612.1 | - | 2 - 3 | 185 | 96 | ||
| FQ224095.1 | - | 2 - 3 | 196 | 96 | ||
| FQ224271.1 | - | 2 - 3 | 197 | 96 | ||
| FQ224320.1 | - | 2 - 3 | 195 | 96 | ||
| FQ224742.1 | - | 2 - 3 | 196 | 96 | ||
| FQ226252.1 | - | 2 - 3 | 196 | 96 | ||
| FQ227715.1 | - | 2 - 3 | 197 | 96 | ||
| FQ227806.1 | - | 2 - 3 | 225 | 96 | ||
| FQ228514.1 | - | 2 - 3 | 196 | 96 | ||
| FQ228556.1 | - | 2 - 3 | 196 | 96 | ||
| FQ228624.1 | - | 2 - 3 | 196 | 96 | ||
| FQ228630.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229028.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229093.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229217.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229279.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229346.1 | - | 2 - 3 | 196 | 96 | ||
| FQ229353.1 | - | 2 - 3 | 196 | 96 | ||
| FQ231339.1 | - | 2 - 3 | 194 | 96 | ||
| FQ231778.1 | - | 2 - 3 | 196 | 96 | ||
| FQ232593.1 | - | 2 - 3 | 196 | 96 | ||
| FQ232768.1 | - | 2 - 3 | 197 | 96 | ||
| FQ233320.1 | - | 2 - 3 | 227 | 96 | ||
| FQ233909.1 | - | 2 - 3 | 199 | 96 | ||
| FQ234047.1 | - | 2 - 3 | 186 | 96 | ||
| FQ234186.1 | - | 2 - 3 | 197 | 96 | ||
| FQ234298.1 | - | 2 - 3 | 196 | 96 | ||
| FQ234781.1 | - | 2 - 3 | 196 | 96 | ||
| U20525.1 | - | 2 - 3 | 168 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tpt1
Genomic Map