Assay Details
| Gene Symbol: NEWGENE_1305281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233223.2 | XP_006233285.1 | 3 - 4 | 500 | 119 | |
| XM_006233224.2 | XP_006233286.1 | 3 - 4 | 615 | 119 | ||
| XM_008761473.1 | XP_008759695.1 | 3 - 4 | 371 | 119 | ||
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106467.3 | NP_001099937.1 | 368 | 119 | ||
| GenBank mRNA | BC160912.1 | - | 357 | 7456962 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_006233224.2 transcripts.
This assay is in the 5'UTR region of the XM_006233224.2 transcripts.
Set Membership: |
|

NEWGENE_1305281
Genomic Map