Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012583.2 | NP_036715.1 | 6 - 7 | 550 | 65 | |
| GenBank mRNA | BC098629.1 | - | 6 - 7 | 550 | 7386548 | 65 |
| FQ212770.1 | - | 134 | 65 | |||
| FQ212939.1 | - | 134 | 65 | |||
| FQ213155.1 | - | 134 | 65 | |||
| FQ215291.1 | - | 134 | 65 | |||
| FQ215779.1 | - | 134 | 65 | |||
| FQ216318.1 | - | 134 | 65 | |||
| FQ219974.1 | - | 134 | 65 | |||
| FQ220248.1 | - | 134 | 65 | |||
| FQ229708.1 | - | 134 | 65 | |||
| FQ230307.1 | - | 134 | 65 | |||
| FQ234297.1 | - | 134 | 65 | |||
| M63983.1 | - | 6 - 7 | 582 | 65 | ||
| M86443.1 | - | 310 | 65 | |||
| S79292.1 | - | 6 - 7 | 582 | 65 | ||
| X62085.1 | - | 510 | 65 | |||
| Gene Symbol: LOC103689983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773659.1 | XP_008771881.1 | 6 - 7 | 626 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hprt1
Genomic Map