Assay Details
Gene Symbol: Hprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012583.2 | NP_036715.1 | 7 - 8 | 597 | 86 | |
GenBank mRNA | BC098629.1 | - | 7 - 8 | 597 | 7386548 | 86 |
FQ212770.1 | - | 181 | 86 | |||
FQ212939.1 | - | 181 | 86 | |||
FQ213155.1 | - | 181 | 86 | |||
FQ215291.1 | - | 181 | 86 | |||
FQ215779.1 | - | 181 | 86 | |||
FQ216318.1 | - | 181 | 86 | |||
FQ219974.1 | - | 181 | 86 | |||
FQ220248.1 | - | 181 | 86 | |||
FQ229708.1 | - | 181 | 86 | |||
FQ230307.1 | - | 181 | 86 | |||
FQ234297.1 | - | 181 | 86 | |||
M63983.1 | - | 7 - 8 | 629 | 86 | ||
M86443.1 | - | 357 | 86 | |||
S79292.1 | - | 7 - 8 | 629 | 86 | ||
X62085.1 | - | 557 | 86 |
Gene Symbol: LOC103689983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773659.1 | XP_008771881.1 | 7 - 8 | 673 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
