Assay Details
| Gene Symbol: Ddx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107824.2 | NP_001101294.2 | 18 - 19 | 1935 | 69 | |
| XM_006233799.2 | XP_006233861.1 | 17 - 18 | 2107 | 69 | ||
| GenBank mRNA | BC160917.1 | - | 18 - 19 | 1935 | 7457829 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P513465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40140 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P513465 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P513465 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P513465 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P513465 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65883 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10834891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834887 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834878 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834880 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834879 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10834875 | - | - |
| Rat Genome 230 2.0 Array | 1374506_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375599_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374506_at | - | - |
| Rat Genome 230 2.0 Array | 1375599_at | - | - |
| Affymetrix HT Rat Focus Array | 1374506_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374506_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300001716.rn.1 | TC0300000175.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300001717.rn.1 | TC0300000175.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375599_at | - | - |
| Rat Expression Set 230 | 1375599_PM_at | - | - |
| Rat Expression Set 230 | 1374506_at | - | - |
| Rat Expression Set 230 | 1374506_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10834875 | - | - |
| Rat Genome U34 Set | rc_AI136398_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21269934 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353289 | - | - |

Ddx31
Genomic Map