Assay Details
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271342.1 | NP_001258271.1 | 6 - 7 | 2810 | 61 | |
XM_006254228.2 | XP_006254290.1 | 6 - 7 | 2810 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
