Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 1 - 2 | 214 | 63 | |
XM_006248558.2 | XP_006248620.1 | 1 - 2 | 232 | 63 | ||
GenBank mRNA | CO401508.1 | - | 1 - 2 | 203 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
