Assay Details
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276711.1 | NP_001263640.1 | 13 - 14 | 1561 | 67 | |
XM_006233358.2 | XP_006233420.1 | 11 - 12 | 1217 | 67 | ||
XM_006233360.2 | XP_006233422.1 | 11 - 12 | 1217 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
