Assay Details
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106225.1 | NP_001099695.1 | 10 - 11 | 1213 | 111 | |
XM_006228216.2 | XP_006228278.1 | 10 - 11 | 1237 | 111 | ||
GenBank mRNA | BC168158.1 | - | 10 - 11 | 1208 | 8364284 | 111 |
FQ212233.1 | - | 7 - 8 | 678 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P482700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174840 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P482700 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P482700 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P482700 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P482700 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43431 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10719028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719035 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719024 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719022 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719025 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719023 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719027 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719032 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10719021 | - | - |
Rat Genome 230 2.0 Array | 1398578_at | - | - |
Rat Genome 230 2.0 Array | 1375905_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397511_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375905_at | - | - |
Rat Genome 230 2.0 Array | 1398578_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397511_at | - | - |
Rat Genome 230 2.0 Array | 1383858_at | - | - |
Rat Genome 230 2.0 Array | 1383858_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375905_at | - | - |
Affymetrix HT Rat Focus Array | 1397511_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383858_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1397511_at | - | - |
Affymetrix HT Rat Focus Array | 1375905_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383858_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100038003.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037991.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100038005.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037996.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037997.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037998.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100038000.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037992.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037995.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037999.rn.1 | TC0100004816.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100037994.rn.1 | TC0100004816.rn.1 | RTA TC |
Rat Expression Set 230 | 1375905_PM_at | - | - |
Rat Expression Set 230 | 1375905_at | - | - |
Rat Expression Set 230 | 1398578_PM_at | - | - |
Rat Expression Set 230 | 1398578_at | - | - |
Rat Expression Set 230 | 1397511_PM_at | - | - |
Rat Expression Set 230 | 1383858_PM_at | - | - |
Rat Expression Set 230 | 1397511_at | - | - |
Rat Expression Set 230 | 1383858_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10719021 | - | - |
Rat Genome U34 Set | rc_AA899079_at | - | - |
Rat Genome U34 Set | rc_AI227624_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20811635 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359698 | - | - |
