Assay Details
| Gene Symbol: Lilrb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228238.2 | XP_006228300.1 | 6 - 7 | 893 | 77 | |
| XM_006228239.2 | XP_006228301.1 | 6 - 7 | 893 | 77 | ||
| GenBank mRNA | AY359280.1 | - | 6 - 7 | 822 | 77 | |
More Information
Set Membership: |
|

Lilrb4
Genomic Map