Assay Details
| Gene Symbol: Cab39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106924.1 | NP_001100394.1 | 6 - 7 | 993 | 147 | |
| XM_006245373.2 | XP_006245435.1 | 7 - 8 | 1142 | 147 | ||
| XM_006245374.2 | XP_006245436.1 | 7 - 8 | 1285 | 147 | ||
| XM_006245375.2 | XP_006245437.1 | 7 - 8 | 1044 | 147 | ||
| GenBank mRNA | FQ225617.1 | - | 3 - 4 | 309 | 147 | |
| FQ226825.1 | - | 2 - 3 | 135 | 147 | ||
| FQ232683.1 | - | 3 - 4 | 309 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cab39
Genomic Map