Assay Details
| Gene Symbol: Ccl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019233.1 | NP_062106.1 | 2 - 3 | 246 | 65 | |
| XM_006245164.2 | XP_006245226.1 | 2 - 3 | 312 | 65 | ||
| GenBank mRNA | AF053312.1 | - | 2 - 3 | 247 | 65 | |
| U90447.1 | - | 2 - 3 | 246 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl20
Genomic Map