Assay Details
| Gene Symbol: Atg16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108809.3 | NP_001102279.2 | 5 - 6 | 645 | 59 | |
| XM_006245419.2 | XP_006245481.1 | 5 - 6 | 823 | 59 | ||
| XM_006245420.2 | XP_006245482.1 | 5 - 6 | 822 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P023541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P804200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P804200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P804200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023541 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P023541 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P804200 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P525665 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1014311 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P804200 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78468 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10925097 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925096 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925094 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925092 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925093 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925099 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925095 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925098 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925102 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10925091 | - | - |
| Rat Genome 230 2.0 Array | 1372375_at | - | - |
| Rat Genome 230 2.0 Array | 1372375_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372375_at | - | - |
| Affymetrix HT Rat Focus Array | 1372375_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900009257.rn.1 | TC0900001084.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372375_PM_at | - | - |
| Rat Expression Set 230 | 1372375_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10925091 | - | - |
| Rat Genome U34 Set | rc_AA892083_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21346393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366066 | - | - |

Atg16l1
Genomic Map