Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004252.1 | NP_001004252.1 | 10 - 11 | 908 | 77 | |
| XM_006245189.2 | XP_006245251.1 | 10 - 11 | 944 | 77 | ||
| GenBank mRNA | BC079364.1 | - | 10 - 11 | 908 | 7109296 | 77 |
| FQ233375.1 | - | 3 - 4 | 197 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farsb
Genomic Map