Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127529.2 | NP_001121001.1 | 15 - 16 | 1752 | 63 | |
| XM_006245177.2 | XP_006245239.1 | 15 - 16 | 1910 | 63 | ||
| XM_006245179.2 | XP_006245241.1 | 15 - 16 | 1703 | 63 | ||
| XM_006245180.2 | XP_006245242.1 | 15 - 16 | 1715 | 63 | ||
| XM_006245182.2 | XP_006245244.1 | 15 - 16 | 1601 | 63 | ||
| XM_008767223.1 | XP_008765445.1 | 14 - 15 | 2808 | 63 | ||
| XM_008767224.1 | XP_008765446.1 | 15 - 16 | 2325 | 63 | ||
| XM_008767225.1 | XP_008765447.1 | 12 - 13 | 2538 | 63 | ||
| GenBank mRNA | BC161869.1 | - | 15 - 16 | 1752 | 7109779 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l
Genomic Map