Assay Details
Gene Symbol: Mrpl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031650.1 | NP_001026820.1 | 1 - 2 | 192 | 71 | |
GenBank mRNA | BC098715.1 | - | 1 - 2 | 192 | 7324407 | 71 |
FQ215293.1 | - | 1 - 2 | 269 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
