Assay Details
| Gene Symbol: Mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031650.1 | NP_001026820.1 | 2 - 3 | 661 | 75 | |
| GenBank mRNA | BC098715.1 | - | 2 - 3 | 661 | 7324407 | 75 |
| FQ215293.1 | - | 2 - 3 | 738 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl44
Genomic Map