Assay Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108808.1 | NP_001102278.1 | 3 - 4 | 784 | 96 | |
| GenBank mRNA | BC158817.1 | - | 391 | 7374173 | 96 | |
| FQ219916.1 | - | 464 | 96 | |||
| FQ220431.1 | - | 507 | 96 | |||
| FQ226325.1 | - | 476 | 96 | |||
| FQ231706.1 | - | 96 | ||||
| FQ232529.1 | - | 476 | 96 | |||
| FQ233479.1 | - | 412 | 96 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4e2
Genomic Map