Assay Details
Gene Symbol: Vom1r47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008916.1 | NP_001008916.1 | 1 | 843 | 123 | |
GenBank mRNA | AY510316.1 | - | 1 | 843 | 123 | |
AY510317.1 | - | 1 | 843 | 123 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
