Assay Details
Gene Symbol: Ube2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108471.1 | NP_001101941.1 | 3 - 4 | 813 | 59 | |
GenBank mRNA | FQ209516.1 | - | 4 - 5 | 406 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
