Assay Details
Gene Symbol: Plac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108395.1 | NP_001101865.1 | 2 - 3 | 170 | 54 | |
XM_006252630.2 | XP_006252692.1 | 2 - 3 | 419 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
