Assay Details
| Gene Symbol: Zcchc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108394.1 | NP_001101864.1 | 3 - 4 | 627 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P815675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135470 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P815675 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P815675 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P815675 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18422 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49570 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57920 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10789728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10789731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10789730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10789729 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10789727 | - | - |
| Rat Genome 230 2.0 Array | 1378948_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386106_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386106_at | - | - |
| Rat Genome 230 2.0 Array | 1388672_at | - | - |
| Rat Genome 230 2.0 Array | 1378948_at | - | - |
| Rat Genome 230 2.0 Array | 1388672_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388672_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386106_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378948_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378948_at | - | - |
| Affymetrix HT Rat Focus Array | 1386106_at | - | - |
| Affymetrix HT Rat Focus Array | 1388672_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600008069.rn.1 | TC1600001103.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388672_at | - | - |
| Rat Expression Set 230 | 1388672_PM_at | - | - |
| Rat Expression Set 230 | 1378948_PM_at | - | - |
| Rat Expression Set 230 | 1386106_PM_at | - | - |
| Rat Expression Set 230 | 1386106_at | - | - |
| Rat Expression Set 230 | 1378948_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10789727 | - | - |
| Rat Genome U34 Set | rc_AI101373_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21045472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373340 | - | - |

Zcchc24
Genomic Map