Assay Details
| Gene Symbol: Dnah12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252671.2 | XP_006252733.1 | 17 - 18 | 2321 | 58 | |
| XM_006252672.2 | XP_006252734.1 | 16 - 17 | 2195 | 58 | ||
| XM_006252673.2 | XP_006252735.1 | 16 - 17 | 2162 | 58 | ||
| XM_006252674.2 | XP_006252736.1 | 17 - 18 | 2321 | 58 | ||
| XM_008771017.1 | XP_008769239.1 | 17 - 18 | 4124 | 58 | ||
| XM_008771018.1 | XP_008769240.1 | 7 - 8 | 936 | 58 | ||
| XM_008771019.1 | XP_008769241.1 | 17 - 18 | 2321 | 58 | ||
| XM_008771020.1 | XP_008769242.1 | 17 - 18 | 2321 | 58 | ||
| XM_008771021.1 | XP_008769243.1 | 17 - 18 | 2321 | 58 | ||
| XM_008771022.1 | XP_008769244.1 | 17 - 18 | 2321 | 58 | ||
| GenBank mRNA | AY032856.1 | - | 2 - 3 | 309 | 58 | |
More Information
Set Membership: |
|

Dnah12
Genomic Map