Assay Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170749.1 | NP_001164220.1 | 8 - 9 | 1565 | 72 | |
| XM_008771008.1 | XP_008769230.1 | 9 - 10 | 1669 | 72 | ||
| XM_008771009.1 | XP_008769231.1 | 3 - 4 | 979 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Asb14
Genomic Map