Assay Details
| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108393.1 | NP_001101863.1 | 18 - 19 | 2887 | 67 | |
| XM_006252537.2 | XP_006252599.1 | 17 - 18 | 15398 | 67 | ||
| XM_006252538.2 | XP_006252600.1 | 18 - 19 | 2934 | 67 | ||
| XM_006252539.2 | XP_006252601.1 | 18 - 19 | 2952 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zmiz1
Genomic Map