Assay Details
| Gene Symbol: Gpr112l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773667.1 | XP_008771889.1 | 20 - 21 | 8541 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr112l| Gene Symbol: Gpr112l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773667.1 | XP_008771889.1 | 20 - 21 | 8541 | 116 | |
Set Membership: |
|