Assay Details
| Gene Symbol: Polr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108886.1 | NP_001102356.1 | 1 - 2 | 131 | 77 | |
| GenBank mRNA | FQ228021.1 | - | 1 - 2 | 102 | 77 | |
| FQ233353.1 | - | 1 - 2 | 98 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr2d
Genomic Map